3-Fluoro-4-(trifluoromethyl)phenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC9216-1G
76627-622EA 21 USD
76627-622
3-Fluoro-4-(trifluoromethyl)phenylacetonitrile 98%
3-Fluoro-4-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
Storage Temperature: Ambient
MDL Number: MFCD00236305
CAS Number: 247113-90-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR