2-Fluoro-4-(trifluoromethyl)phenylacetonitrile 94%

Supplier: APOLLO SCIENTIFIC
Danger

PC4377F-1G PC4377F-10G PC4377F-5G
76626-488EA 285 USD
76626-488 76626-490 76626-492
2-Fluoro-4-(trifluoromethyl)phenylacetonitrile 94%
2-Fluoro-4-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
Storage Temperature: Ambient
MDL Number: MFCD00061179
CAS Number: 239087-11-7
UN: 3276
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR