2-Amino-4-fluoropyridine

Supplier: APOLLO SCIENTIFIC

PC8826-1G
76609-662EA 16 USD
76609-662
2-Amino-4-fluoropyridine
2-Amino-4-fluoropyridine
Formula: C₅H₅FN₂
MW: 112.04 g/mol
MDL Number: MFCD09260906
CAS Number: 944401-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR