2-Fluoro-3-methylbenzaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

PC9906-5G PC9906-1G
76610-632EA 42 USD
76610-632 76610-624
2-Fluoro-3-methylbenzaldehyde
2-Fluoro-3-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138.1390 g/mol
Boiling Pt: 77…79 °C/10mmHg
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD06797948
CAS Number: 886762-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR