3'-(Pentafluorothio)acetophenone 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC51188-5G PC51188-1G
76604-396EA 13 USD
76604-396 76606-276
3'-(Pentafluorothio)acetophenone 97%
3'-(Pentafluorothio)acetophenone
Formula: C₈H₇F₅OS
Storage Temperature: Ambient
MDL Number: MFCD16652407
CAS Number: 159896-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR