2-Bromo-3'-chloro-4'-fluoroacetophenone 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 3-Chloro-4-fluorophenacyl bromide

PC3542-5G PC3542-1G PC3542-25G
76618-210EA 21 USD
76618-210 76618-214 76618-212
2-Bromo-3'-chloro-4'-fluoroacetophenone 98%
2-Bromo-3'-chloro-4'-fluoroacetophenone
Formula: C₈H₅BrClFO
MW: 251.48
Melting Pt: 35…37 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05154991
CAS Number: 63529-30-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR