3-Fluoro-4-formylphenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Fluoro-4-formylbenzeneboronic acid

PC6911-5G PC6911-25G PC6911-100G
76603-872EA 596 USD
76603-872 76603-874 76603-876
3-Fluoro-4-formylphenylboronic acid 98%
3-Fluoro-4-formylphenylboronic acid
Formula: C₇H₆BFO₃
MW: 167.93 g/mol
Melting Pt: 240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093051
CAS Number: 248270-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR