3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Bis(3,5-bis(trifluoromethyl)phenyl)methanone

PC6934-5G PC6934-1G
76607-986EA 110 USD
76607-986 76607-984
3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone 97%
3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone
Formula: C₁₇H₆F₁₂O
MW: 454.22 g/mol
Melting Pt: 139…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00042474
CAS Number: 175136-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR