4-(Trifluoromethylthio)benzylamine 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC9326-1G
PC9326-5G
PC9326-25G
76627-642EA
101
USD
76627-642
76627-646
76627-644
4-(Trifluoromethylthio)benzylamine 98%
4-(Trifluoromethylthio)benzylamine
Formula:
C₈H₈F₃NS Boiling Pt: 224…225 °C Density: 1.31 g/cm³ (25 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190129 CAS Number: 128273-56-3 UN: 3267 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...