α,4-Dibromo-3-fluorotoluene 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 4-Bromo-3-fluorobenzyl bromide1-Bromo-4-(bromomethyl)-2-fluorobenzene

PC5721-25G PC5721-1G PC5721-5G
76614-328EA 13 USD
76614-328 76614-332 76614-330
α,4-Dibromo-3-fluorotoluene 97%
α,4-Dibromo-3-fluorotoluene
Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Boiling Pt: 112…114 °C/8mmHg
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD11847179
CAS Number: 127425-73-4
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR