α,3-Dibromo-2-fluorotoluene

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Bromo-2-fluorobenzyl bromide1-Bromo-3-(bromomethyl)-2-fluorobenzene

PC3168-5G PC3168-1G PC3168-25G
76623-278EA 17 USD
76623-278 76623-282 76623-280
α,3-Dibromo-2-fluorotoluene
α,3-Dibromo-2-fluorotoluene
Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Boiling Pt: 60 °C/0.1mmHg
Storage Temperature: Ambient
MDL Number: MFCD09842442
CAS Number: 149947-16-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR