1,5-Dibromo-2,4-difluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC9091-100G PC9091-25G PC9091-5G
76610-044EA 18 USD
76610-044 76610-042 76610-040
1,5-Dibromo-2,4-difluorobenzene 98%
1,5-Dibromo-2,4-difluorobenzene
Formula: C₆H₂Br₂F₂
MW: 271.8849 g/mol
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD08458122
CAS Number: 28342-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR