1,3-Dibromo-2,5-difluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC56083-1G PC56083-25G PC56083-5G
76602-238EA 33 USD
76602-238 76602-236 76602-234
1,3-Dibromo-2,5-difluorobenzene 99%
1,3-Dibromo-2,5-difluorobenzene
Formula: C₆H₂Br₂F₂
MW: 271.8900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878120
CAS Number: 128259-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR