4'-Fluoro-3'-methoxyacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(4-Fluoro-3-methoxyphenyl)ethanone

PC7322-10G PC7322-1G PC7322-5G
76608-376EA 116 USD
76608-376 76608-374 76608-372
4'-Fluoro-3'-methoxyacetophenone 98%
4'-Fluoro-3'-methoxyacetophenone
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Melting Pt: 51…53 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00272135
CAS Number: 64287-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR