3'-Fluoro-2'-hydroxyacetophenone 98%

Supplier: APOLLO SCIENTIFIC

Warning

PC49371-5G PC49371-250MG PC49371-1G
76607-292EA 209 USD
76607-292 76605-768 76604-596
3'-Fluoro-2'-hydroxyacetophenone 98%
3'-Fluoro-2'-hydroxyacetophenone
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD09055140
CAS Number: 699-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR