2-Bromo-3',5'-difluoroacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-1-(3,5-difluorophenyl)ethanone3,5-Difluorophenacyl bromide

PC4473-5G PC4473-1G PC4473-25G
76618-360EA 48 USD
76618-360 76618-364 76618-362
2-Bromo-3',5'-difluoroacetophenone 98%
2-Bromo-3',5'-difluoroacetophenone
Formula: C₈H₅BrF₂O
MW: 235.0255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094245
CAS Number: 220607-75-0
UN: 2920
ADR: 8+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR