4-(Difluoromethoxy)benzylamine 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: (4-(Difluoromethoxy)phenyl)methanamine

PC4305-25G PC4305-1G PC4305-5G
76613-910EA 230 USD
76613-910 76613-912 76613-908
4-(Difluoromethoxy)benzylamine 98%
4-(Difluoromethoxy)benzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236231
CAS Number: 177842-14-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR