3-(Difluoromethoxy)benzylamine 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: (3-(Difluoromethoxy)phenyl)methanamine

PC8902-5G PC8902-1G
76614-680EA 48 USD
76614-680 76614-682
3-(Difluoromethoxy)benzylamine 98%
3-(Difluoromethoxy)benzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236230
CAS Number: 244022-71-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR