2-(Difluoromethoxy)benzylamine 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: (2-(Difluoromethoxy)phenyl)methanamine

PC7845-1G PC7845-5G PC7845-25G
76614-614EA 20 USD
76614-614 76614-618 76614-616
2-(Difluoromethoxy)benzylamine 98%
2-(Difluoromethoxy)benzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236229
CAS Number: 243863-36-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR