4-Fluoro-2-methylphenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Methyl-4-fluorophenylboronic acid4-Fluoro-2-methylbenzeneboronic acid

PC6913-25G PC6913-100G PC6913-5G
76603-886EA 88 USD
76603-886 76603-888
4-Fluoro-2-methylphenylboronic acid 98%
4-Fluoro-2-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 191…196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093072
CAS Number: 139911-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR