2-Bromo-2'-fluoro-4'-(trifluoromethyl)acetophenone

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Fluoro-4-(trifluoromethyl)phenacyl bromide

PC8406-1G PC8406-25G PC8406-5G
76614-644EA 250 USD
76614-644 76614-642 76614-646
2-Bromo-2'-fluoro-4'-(trifluoromethyl)acetophenone
2-Bromo-2'-fluoro-4'-(trifluoromethyl)acetophenone
Formula: C₉H₅BrF₄O
MW: 285.0330 g/mol
Melting Pt: 30…37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094505
CAS Number: 537050-12-7
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR