4-(Trifluoromethylthio)benzyl bromide 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: (4-(Bromomethyl)phenyl)(trifluoromethyl)sulfane

PC7769K-1G PC7769K-100MG
76624-500EA 13 USD
76624-500 76624-502
4-(Trifluoromethylthio)benzyl bromide 97%
4-(Trifluoromethylthio)benzyl bromide
Formula: C₈H₆BrF₃S
MW: 271.10 g/mol
Boiling Pt: 115…118 °C/13mmHg
Melting Pt: 53…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00129189
CAS Number: 21101-63-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR