4'-Nitro-3'-(trifluoromethyl)acetanilide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-(4-Nitro-3-(trifluoromethyl)phenyl)acetamide

PC5296-1G PC5296-5G
76624-286EA 13 USD
76624-286 76624-288
4'-Nitro-3'-(trifluoromethyl)acetanilide 97%
4'-Nitro-3'-(trifluoromethyl)acetanilide
Formula: C₉H₇F₃N₂O₃
MW: 248.1600 g/mol
Melting Pt: 123 °C
Storage Temperature: Ambient
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR