4,5-Difluoro-2-nitrotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,2-Difluoro-4-methyl-5-nitrobenzene

PC4368-25G PC4368-5G PC4368-100G
76618-348EA 13 USD
76618-348 76618-346 76618-344
4,5-Difluoro-2-nitrotoluene 98%
4,5-Difluoro-2-nitrotoluene
Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973772
CAS Number: 127371-50-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR