4-Fluoro-3-(trifluoromethoxy)benzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7418-5G PC7418-1G PC7418-10G
76608-472EA 14 USD
76608-472 76608-470 76608-468
4-Fluoro-3-(trifluoromethoxy)benzoic acid 98%
4-Fluoro-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD06660161
CAS Number: 886496-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR