3-Fluoro-4-iodobenzonitrile

Supplier: APOLLO SCIENTIFIC

Warning

PC9581-5G PC9581-25G
76610-462EA 19 USD
76610-462 76610-460
3-Fluoro-4-iodobenzonitrile
3-Fluoro-4-iodobenzonitrile
Formula: C₇H₃FIN
MW: 247.01 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD07782076
CAS Number: 887266-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR