2,4-Difluoro-5-methoxyaniline 99%

Supplier: APOLLO SCIENTIFIC

Warning

PC7611-5G PC7611-500MG PC7611-1G
76608-672EA 833 USD
76608-672 76608-670 76608-668
2,4-Difluoro-5-methoxyaniline 99%
2,4-Difluoro-5-methoxyaniline
Formula: C₇H₇NOF₂
Storage Temperature: Refrigerator
MDL Number: MFCD08689699
CAS Number: 98446-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR