3-Hydroxy-5-(trifluoromethyl)benzoic acid

Supplier: APOLLO SCIENTIFIC

Warning

PC9912-10G PC9912-5G PC9912-1G
76610-644EA 47 USD
76610-644 76610-642 76610-646
3-Hydroxy-5-(trifluoromethyl)benzoic acid
3-Hydroxy-5-(trifluoromethyl)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 191…193 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07368783
CAS Number: 328-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR