3,4,5,6-Tetrafluorosalicylic acid

Supplier: APOLLO SCIENTIFIC

Warning

PC52121-5G PC52121-1G
76600-324EA 446 USD
76600-324 76600-322
3,4,5,6-Tetrafluorosalicylic acid
3,4,5,6-Tetrafluorosalicylic acid
Formula: C₇H₂F₄O₃
Melting Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00483242
CAS Number: 14742-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR