4-Nitrophenylsulfur pentafluoride

Supplier: APOLLO SCIENTIFIC

Danger

PC8766-1G PC8766-5G PC8766-25G
76619-006EA 43 USD
76619-006 76619-008 76619-010
4-Nitrophenylsulfur pentafluoride
4-Nitrophenylsulphur pentafluoride
Formula: C₆H₄F₅NO₂S
MW: 249.16 g/mol
Boiling Pt: 76…77 °C/12mmHg
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00221619
CAS Number: 2613-27-6
EINECS: 447-610-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR