5-Chloro-2-fluorophenylhydrazine hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (5-Chloro-2-fluorophenyl)hydrazine hydrochloride

PC7116-1G PC7116-5G PC7116-25G
76608-136EA 13 USD
76608-136 76608-140 76608-138
5-Chloro-2-fluorophenylhydrazine hydrochloride 97%
5-Chloro-2-fluorophenylhydrazine hydrochloride
Formula: C₆H₇Cl₂FN₂
MW: 197.0376 g/mol
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD03094414
CAS Number: 529512-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR