3-Chloro-2-fluorophenylhydrazine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3-Chloro-2-fluorophenyl)hydrazine hydrochloride

PC5302-25G PC5302-5G
76600-766EA 70 USD
76600-766 76600-752
3-Chloro-2-fluorophenylhydrazine hydrochloride
3-Chloro-2-fluorophenylhydrazine hydrochloride
Formula: C₆H₇Cl₂FN₂
MW: 197.0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094280
CAS Number: 517920-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR