Potassium benzyltrifluoroborate 97%

Supplier: APOLLO SCIENTIFIC

Warning

PC50055-5G
76605-698EA 98 USD
76605-698
Potassium benzyltrifluoroborate 97%
Potassium benzyltrifluoroborate
Formula: C₇H₇BF₃K
MW: 198.0350 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03427261
CAS Number: 329976-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR