TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: TATU

PC902639-25G PC902639-100G PC902639-5G
76614-688EA 21 USD
76614-688 76614-686 76614-684
TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 98%
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Formula: C₁₀H₁₅BF₄N₆O
MW: 322.1 g/mol
Melting Pt: 184 °C (dec.)
Storage Temperature: Ambient
CAS Number: 873798-09-5
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR