Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

PC8598-100MG PC8598-1G
76609-460EA 98 USD
76609-460 76609-458
Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Methyl 5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Formula: C₁₁H₇F₃O₂S
MW: 260.2323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03012932
CAS Number: 146137-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR