3-(Trifluoromethyl)cyclohexanone 97%

Supplier: APOLLO SCIENTIFIC

Warning

PC7668M-1G PC7668M-5G
76608-718EA 315 USD
76608-718 76608-716
3-(Trifluoromethyl)cyclohexanone 97%
3-(Trifluoromethyl)cyclohexanone
Formula: C₇H₉F₃O
MW: 166.14
Boiling Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00034662
CAS Number: 585-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR