4-(1,1,2,2-Tetrafluoroethoxy)toluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC6780-25G PC6780-50G
76618-874EA 21 USD
76618-874 76618-872
4-(1,1,2,2-Tetrafluoroethoxy)toluene 98%
4-(1,1,2,2-Tetrafluoroethoxy)toluene
Formula: C₉H₈F₄O
MW: 208.1528 g/mol
Boiling Pt: 67…69 °C/14mmHg
Density: 1.25 g/cm³ (25 °C)
Flash Pt: 61.5 °C
Storage Temperature: Ambient
EINECS: 217-086-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR