(3-(Trifluoromethyl)phenoxy)acetonitrile 97%

Supplier: APOLLO SCIENTIFIC

Warning

PC7696-10G PC7696-1G PC7696-5G
76627-498EA 43 USD
76627-498 76627-496 76627-494
(3-(Trifluoromethyl)phenoxy)acetonitrile 97%
(3-(Trifluoromethyl)phenoxy)acetonitrile
Formula: C₉H₆F₃NO
Boiling Pt: 82…84 °C/0.5mmHg
Storage Temperature: Ambient
CAS Number: 2145-31-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR