4-Chloro-2-fluorophenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 4-Chloro-2-fluorobenzeneacetonitrile

PC9472-100G PC9472-25G PC9472-5G
76627-652EA 14 USD
76627-652 76627-650 76627-648
4-Chloro-2-fluorophenylacetonitrile 98%
4-Chloro-2-fluorophenylacetonitrile
Formula: C₈H₅ClFN
MW: 169.59 g/mol
Melting Pt: 36…39 °C
Storage Temperature: Ambient
MDL Number: MFCD01312758
CAS Number: 75279-53-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR