4-Fluoro-3-methoxyphenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Fluoro-3-methoxybenzeneboronic acid(4-Fluoro-3-methoxyphenyl)boronic acid

PC5053-5G PC5053-250MG PC5053-1G
76604-680EA 69 USD
76604-680 76606-672 76606-576
4-Fluoro-3-methoxyphenylboronic acid 98%
4-Fluoro-3-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056358
CAS Number: 854778-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR