Methyl-3,4-difluorobenzoate 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Methyl 3,4-difluorobenzoate
PC5102-25G
PC5102-100G
76606-592EA
43
USD
76606-592
76606-202
Methyl-3,4-difluorobenzoate 97%
Methyl 3,4-difluorobenzoate
Formula:
C₈H₆F₂O₂ MW: 172.13 g/mol Boiling Pt: 87…89 °C/15mmHg Melting Pt: 23…27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00276597 CAS Number: 369-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...