Lithium trifluoromethanesulfonate 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC5012-25G
PC5012-100G
76605-900EA
16
USD
76605-900
76604-338
Lithium trifluoromethanesulfonate 97%
Lithium trifluoromethanesulphonate
Formula:
CF₃LiO₃S Melting Pt: 423 °C Density: 1.9 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013227 CAS Number: 33454-82-9 EINECS: 251-528-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...