4'-Hydroxy-3'-(trifluoromethyl)acetophenone

Supplier: APOLLO SCIENTIFIC

Warning

PC6845-1G PC6845-250MG
76603-840EA 25 USD
76603-840 76603-838
4'-Hydroxy-3'-(trifluoromethyl)acetophenone
4'-Hydroxy-3'-(trifluoromethyl)acetophenone
Formula: C₉H₇F₃O₂
Melting Pt: 174…179 °C
Storage Temperature: Ambient
MDL Number: MFCD01091005
CAS Number: 149105-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR