(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: (2R)-(+)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

PC5063C-5G PC5063C-1G
76605-230EA 307 USD
76605-230 76606-896
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

For determination of optical purities of alcohols or amines.

Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Boiling Pt: 111 °C/1mmHg
Melting Pt: 43…45 °C
Density: 1.344 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004184
CAS Number: 20445-31-2
EINECS: 243-829-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR