Perfluoroheptanoic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Perfluoroenanthic acidTridecafluoroheptylic acidTridecafluoroenanthic acidTridecafluoroheptanoic acid

PC6034-25G PC6034-100G
76624-434EA 137 USD
76624-434 76624-436
Perfluoroheptanoic acid 98%
Perfluoroheptanoic acid
Formula: C₇HF₁₃O₂
Boiling Pt: 175 °C
Melting Pt: 30 °C
Density: 1.792 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00039604
CAS Number: 375-85-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR