4-Fluoro-3-(trifluoromethoxy)phenol

Supplier: APOLLO SCIENTIFIC
Warning

PC500166-1G PC500166-5G
76606-810EA 133 USD
76606-810 76605-026
4-Fluoro-3-(trifluoromethoxy)phenol
4-Fluoro-3-(trifluoromethoxy)phenol
Formula: C₇H₄F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06660257
CAS Number: 886501-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR