2-Fluoro-5-iodobenzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC9095-1G PC9095-5G PC9095-25G
76619-028EA 47 USD
76619-028 76619-030 76619-026
2-Fluoro-5-iodobenzaldehyde 97%
2-Fluoro-5-iodobenzaldehyde
Formula: C₇H₄FIO
MW: 250.01 g/mol
Melting Pt: 41…46 °C
Storage Temperature: Ambient
MDL Number: MFCD03093999
CAS Number: 146137-76-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR