Ethyl-4,4,4-trifluoroacetoacetate 90%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl trifluoroacetoacetateEthyl 4,4,4-trifluoro-3-oxobutanoate

PC49444-100G PC49444-1KG PC49444-500G
76626-612EA 29 USD
76626-612 76626-610 76626-608
Ethyl-4,4,4-trifluoroacetoacetate 90%
Ethyl 4,4,4-trifluoroacetoacetate
Formula: C₆H₇F₃O₃
MW: 184.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000424
CAS Number: 372-31-6
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR