2'-Fluoro-4'-methylacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(2-Fluoro-4-methylphenyl)ethanone

PC5942-5G PC5942-1G
76603-372EA 31 USD
76603-372 76603-374
2'-Fluoro-4'-methylacetophenone
2'-Fluoro-4'-methylacetophenone
Formula: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094331
CAS Number: 29427-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR