2,3-Difluoro-4-methoxybenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7137-1G
76627-380EA 57 USD
76627-380
2,3-Difluoro-4-methoxybenzonitrile 98%
2,3-Difluoro-4-methoxybenzonitrile
Formula: C₈H₅F₂NO
MW: 169.13 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD03428596
CAS Number: 256417-12-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR